Iron(III) oxide ≥99.9%, powder
Supplier: Thermo Scientific Chemicals
Synonyms:
Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide
047247-18
047247-30
047247-A1
AA47247-18EA
78.14
USD
AA47247-18
AA47247-30
AA47247-A1
Iron(III) oxide ≥99.9%, powder
Iron (III) oxide
Iron(III) oxide, 99.9% (metals basis)
Purity: 99.9%
CAS No.: 1309-37-1
Molecular Formula: Fe2O3
Formula Weight: 159.69
Physical Form: Powder
Odor: Odorless
Melting Point: 1565°
MDL No.: MFCD00011008
Formula:
Fe₂O₃ Melting Pt: 1565 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011008 CAS Number: 1309-37-1 EINECS: 215-168-2 Merck Index: 14,4028 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...