Ytterbium ≥99.9%, ingot

Supplier: Thermo Scientific Chemicals

047227-KF
AA47227-KFEA 587 USD
AA47227-KF
Ytterbium ≥99.9%, ingot
Ytterbium

~40g/piece, Packaged in air stable poly bottle

Formula: Yb
Storage Temperature: Ambient
MDL Number: MFCD00011286
CAS Number: 7440-64-4
EINECS: 231-173-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR