Aluminium oxide, γ-phase ≥99.997% (metals basis), powder

Supplier: Thermo Scientific Chemicals

047202-14 047202-22 047202-36
AA47202-14EA 57.3 USD
AA47202-14 AA47202-22 AA47202-36
Aluminium oxide, γ-phase ≥99.997% (metals basis), powder
Aluminium oxide, γ-phase

Aluminum oxide, ^y-phase, 99.997% (metals basis). 2.5 Micron Powder, S.A. 110-125m^2/g.


Purity: 99.997% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 2.5 Micron Powder, S.A. 110-125m2/g Odor: Odorless MDL No.: MFCD00003424

Formula: Al₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now

Specification Test Results

Assay Minimum 99.997


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR