Tellurium, AAS Standard Solution, Specpure®, Te 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
047130-AE
AA47130-AEEA
33.5
USD
AA47130-AE
AA47130-AP
Tellurium, AAS Standard Solution, Specpure®, Te 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Tellurium Standards
Tellurium, AAS standard solution, Specpure|r, Te 1000^mg/ml
Molecular Formula: Te in 30% HCl
Physical Form: Liquid
Odor: Acidic
UN No.: 1789
MDL No.: MFCD00134062
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...