Sodium hydroxide 3 mol/l (3 N) in aqueous solution analytical reagent
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
047124-K2
047124-K7
AA47124-K2EA
89.18
USD
AA47124-K2
AA47124-K7
Sodium hydroxide 3 mol/l (3 N) in aqueous solution analytical reagent
Sodium hydroxide
Sodium hydroxide, 3.0N Standardized Solution
CAS No.: 1310-73-2
Molecular Formula: NaOH
Formula Weight: 40.00
Physical Form: Liquid
Appearance: Colorless
UN No.: 1824
MDL No.: MFCD00003548
Formula:
NaOH Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...