Silicon dioxide ≥99.95% (metals basis) -40 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
047098-14
047098-22
AA47098-14EA
58.88
USD
AA47098-14
AA47098-22
Silicon dioxide ≥99.95% (metals basis) -40 mesh
Silicium dioxide
Silicon(IV) oxide, 99.95% (metals basis), -40 Mesh Powder.
Purity: 99.95%
CAS No.: 7631-86-9
Molecular Formula: SiO2
Formula Weight: 60.09
Physical Form: -40 Mesh Powder
Density: 2.2
Appearance: White
Odor: Odorless
Melting Point: 1710°
Boiling Point: 2230°
MDL No.: MFCD00011232
Formula:
SiO₂ Boiling Pt: 2230 °C Melting Pt: 1710 °C Density: 2.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 7631-86-9 EINECS: 231-545-4 Merck Index: 14,8493 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...