Platinum, expanded metal mesh, gauze, Unimesh N7433, Thickness 0.34 mm (0.013 in)

Supplier: Thermo Scientific Chemicals

046966-FF 046966-FI
AA46966-FFEA 429.38 USD
AA46966-FF AA46966-FI
Platinum, expanded metal mesh, gauze, Unimesh N7433, Thickness 0.34 mm (0.013 in)
Platinum

  • ~1.0g/25x25mm, Mesh


CAS No.: 7440-06-4 Physical Form: ≈1.0g/25x25mm, Mesh Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR