Molybdenum ≥99.95% (metals basis), foil, Thickness 0.05 mm (0.002 in)

Supplier: Thermo Scientific Chemicals

046943-GH 046943-HW 046943-HX
AA46943-GHEA 72.1 USD
AA46943-GH AA46943-HW AA46943-HX
Molybdenum ≥99.95% (metals basis), foil, Thickness 0.05 mm (0.002 in)
Molybdenum

  • ~5.1g/100x100mm


Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈5.1g/100x100mm MDL No.: MFCD00003465

Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now

Specification Test Results

Assay Minimum 99.95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR