Cerium(IV) oxide 25% in water, colloidal dispersion, NanoArc® CE-6440

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ceric oxide, Cerium dioxide

046763-22 046763-36
AA46763-22EA 58 USD
AA46763-22 AA46763-36
Cerium(IV) oxide 25% in water, colloidal dispersion, NanoArc® CE-6440
Cerium (IV) oxide

30nm APS for dry powder (typical)


Suitable for use in ultrafine acid side polishing slurries


Suitable for use in ultrafine acid side polishing slurries

Formula: CeO₂
Storage Temperature: Ambient
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR