Hastelloy C® alloy, gauze woven from wire 40 mesh Ø 0.19 mm (0.0075 in)
Supplier: Thermo Scientific Chemicals
046690-FL 046690-GT 046690-HB
AA46690-FLEA 38.7 USD
AA46690-FL AA46690-GT AA46690-HB
Hastelloy C® alloy, gauze woven from wire 40 mesh Ø 0.19 mm (0.0075 in)
Hastelloy C® alloy
Wire Cloth
Physical Form: Wire Cloth MDL No.: MFCD00801065