Indium ≥99.998% (metals basis), wire, Puratronic®, annealed, Diameter 1.0 mm (0.04 in)
Supplier: Thermo Scientific Chemicals
046661-G1
046661-G5
046661-G9
AA46661-G1EA
112
USD
AA46661-G1
AA46661-G5
AA46661-G9
Indium ≥99.998% (metals basis), wire, Puratronic®, annealed, Diameter 1.0 mm (0.04 in)
Indium
- ~5.74g/m
Purity: 99.998+%
CAS No.: 7440-74-6
Physical Form: ≈5.74g/m
Odor: Odorless
MDL No.: MFCD00134048
Formula:
In MW: 114.82 g/mol Boiling Pt: 2000 Melting Pt: 156 Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Specification Test Results
Assay Minimum | 99.998 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...