Silicon ≥99.999% (metals basis), powder -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

046654-09 046654-18
AA46654-09EA 88.5 USD
AA46654-09 AA46654-18
Silicon ≥99.999% (metals basis), powder -325 mesh
Silicon

Purity: 99.999% CAS No.: 7440-21-3 Odor: Odorless UN No.: 1346 MDL No.: MFCD00085311

Formula: Si
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR