Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 100 mesh Ø 0.025 mm (0.001 in) Type 304
Supplier: Thermo Scientific Chemicals
|
Danger
|
046628-FL
046628-GT
046628-HB
AA46628-FLEA
26.7
USD
AA46628-FL
AA46628-GT
AA46628-HB
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 100 mesh Ø 0.025 mm (0.001 in) Type 304
Stainless steel
Wire Cloth
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Physical Form: Wire Cloth
Odor: Odorless
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...