Copper, gauze woven from wire 140 mesh Ø 0.056 mm (0.0022 in)
Supplier: Thermo Scientific Chemicals
046603-FL
046603-GT
046603-HB
AA46603-FLEA
30.8
USD
AA46603-FL
AA46603-GT
AA46603-HB
Copper, gauze woven from wire 140 mesh Ø 0.056 mm (0.0022 in)
Copper
- Wire Cloth
CAS No.: 7440-50-8
Physical Form: Wire Cloth
Odor: Odorless
MDL No.: MFCD00010965
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...