Bismuth(III) oxide ≥99.9%, nanopowder
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bismuth Trioxide, Dibismuth trioxide
046314-14
046314-22
046314-36
AA46314-14EA
31.37
USD
AA46314-14
AA46314-22
AA46314-36
Bismuth(III) oxide ≥99.9%, nanopowder
Bismuth (III) oxide
Purity: 99.9%
CAS No.: 1304-76-3
Molecular Formula: Bi2O3
Formula Weight: 465.96
Physical Form: 80-200nm APS Powder, S.A. 3.8-5.5m2/g
Density: 8.9
Appearance: Yellow
Melting Point: 817°
Boiling Point: ca 1890°
Merck Reference: 14,1273
MDL No.: MFCD00003462
Formula:
BiO₃ Boiling Pt: ∼1890 °C Melting Pt: 817 °C Density: 8.9 Storage Temperature: Ambient |
MDL Number:
MFCD00003462 CAS Number: 1304-76-3 EINECS: 215-134-7 Merck Index: 14,1273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...