Platinum 5% on activated carbon wetted with ca. 50% water, powder, reduced Type B105022-5
Supplier: Thermo Scientific Chemicals
CAS No.: 7440-06-4
Physical Form: 900-1100 m²/g
Appearance: Dark grey to black
Odor: Odorless
MDL No.: MFCD00011179
Ordering information: Sold on a dry weight basis. Unit weight excludes water weight.
Caution: Incompatible with acids and oxidizing agents
Formula:
Pt Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...