Dysprosium ≥99.9% (REO, rare earth oxide basis), Powder -200 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
045918-06
045918-14
AA45918-06EA
373
USD
AA45918-06
AA45918-14
Dysprosium ≥99.9% (REO, rare earth oxide basis), Powder -200 mesh
Dysprosium
- Packaged under argon
Purity: 99.9%
CAS No.: 7429-91-6
Physical Form: Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Silver
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00010982
Formula:
Dy Storage Temperature: Ambient |
MDL Number:
MFCD00010982 CAS Number: 7429-91-6 EINECS: 231-073-9 UN: 3089 ADR: 4.1,II |
Specification Test Results
Assay Minimum | 99.9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...