Gadolinium ≥99.9% (REO, rare earth oxide basis), powder < 200 mesh

Supplier: Thermo Scientific Chemicals
Danger

045915-14 045915-06
AA45915-14EA 622 USD
AA45915-14 AA45915-06
Gadolinium ≥99.9% (REO, rare earth oxide basis), powder < 200 mesh
Gadolinium

Purity: 99.9% CAS No.: 7440-54-2 Physical Form: Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Silver Odor: Odorless UN No.: 3089 MDL No.: MFCD00011022

Formula: Gd
MW: 157.25
Density: 7.886 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2
UN: 3089
ADR: 4.1 II

Order Now

Specification Test Results

Total Rare Earth Oxide Impurities: 0.1% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR