Praseodymium ≥99.9% (REO, rare earth oxide basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Danger

045910-06 045910-14
AA45910-06EA 250 USD
AA45910-06 AA45910-14
Praseodymium ≥99.9% (REO, rare earth oxide basis), powder -200 mesh
Praseodymium

  • Packaged under argon


Purity: 99.9% CAS No.: 7440-10-0 Physical Form: Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Silver Odor: Odorless UN No.: 3089 MDL No.: MFCD00011174

Formula: Pr
MW: 140.91
Storage Temperature: Ambient
MDL Number: MFCD00011174
CAS Number: 7440-10-0
EINECS: 231-120-3
UN: 3089
ADR: 4.1 II

Order Now

Specification Test Results

Assay Minimum 99.9


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR