Lanthanum ≥99.9% (REO, rare earth oxide basis), powder < 200 mesh

Supplier: Thermo Scientific Chemicals
Danger

045908-06 045908-14
AA45908-06EA 156 USD
AA45908-06 AA45908-14
Lanthanum ≥99.9% (REO, rare earth oxide basis), powder < 200 mesh
Lanthanum

Purity: 99.9% CAS No.: 7439-91-0 Physical Form: Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Silver Odor: Odorless UN No.: 3089 MDL No.: MFCD00011066

Formula: La
Boiling Pt: 3469 °C (1013 hPa)
Melting Pt: 885 °C
Density: 6.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011066
CAS Number: 7439-91-0
EINECS: 231-099-0
UN: 3089
ADR: 4.1,II
Merck Index: 12,05374

Order Now

Specification Test Results

Form Packaged under argon


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR