Ethanol denatured ≥91.6% (v/v) (denatured with 3.7% MeOH, 1.9% MIBK, 1% each of heptane, ethylacetate and toluene)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethyl alcohol denatured
045844-K2
045844-K7
045844-M1
AA45844-K2EA
36.7
USD
AA45844-K2
AA45844-K7
AA45844-M1
Ethanol denatured ≥91.6% (v/v) (denatured with 3.7% MeOH, 1.9% MIBK, 1% each of heptane, ethylacetate and toluene)
Ethanol denatured
Purity: 91.6%
CAS No.: 64-17-5
Molecular Formula: C2H6O
Formula Weight: 46.08
Flash Point: 8°(46°F)
Physical Form: Liquid
Density: 0.789
Refractive Index: 1.3610
Sensitivity: Hygroscopic
Appearance: Colorless
Melting Point: -130°
Boiling Point: 78°
Beilstein Reference: 1718733
UN No.: 1170
MDL No.: MFCD00003568
Formula:
H₃CCH₂OH MW: 46.07 g/mol Boiling Pt: 78 °C Melting Pt: –130 °C Density: 0.789 Flash Pt: 8 °C (46 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 EINECS: 200-578-6 UN: 1170 ADR: 3 II Merck Index: 14,3760 |
Specification Test Results
Assay Minimum | 91.6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...