Cobalt(II,III) oxide ≥99.7% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tricobalt tetraoxide, Cobaltic tetraoxide

045806-0B 045806-18 045806-30
AA45806-0BEA 917 USD
AA45806-0B AA45806-18 AA45806-30
Cobalt(II,III) oxide ≥99.7% (metals basis)
Cobalt (II,III) oxide

Purity: 99.7% CAS No.: 1308-06-1 Molecular Formula: Co3O4 Formula Weight: 240.80 Physical Form: 2-6 Micron Powder Density: 6.11 Appearance: Black Melting Point: 900° dec. Merck Reference: 14,2429 UN No.: 3077 MDL No.: MFCD00010939

Formula: Co₃O₄
MW: 240.8 g/mol
Melting Pt: 895 (dec.)
Density: 6.11
Storage Temperature: Ambient
MDL Number: MFCD00010939
CAS Number: 1308-06-1
EINECS: 215-157-2
UN: 3077
ADR: 9 III
Merck Index: 14,2429

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR