Sodium hydroxide ≥99.99% (metals basis), beads/pellets
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
045780-06
045780-14
045780-22
AA45780-06EA
31.15
USD
AA45780-06
AA45780-14
AA45780-22
Sodium hydroxide ≥99.99% (metals basis), beads/pellets
Sodium hydroxide
Purity: 99.99%
CAS No.: 1310-73-2
Molecular Formula: NaOH
Formula Weight: 40.00
Density: 2.13
Sensitivity: Air Sensitive and Hygroscopic
Appearance: White to off-white
Odor: Odorless
pH: 14
Melting Point: 316°
Merck Reference: 14,8627
UN No.: 1823
MDL No.: MFCD00003548
Formula:
NaOH Boiling Pt: 1390 °C (1013 hPa) Melting Pt: 316 °C Density: 2.13 Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8,II REACH: 01-2119457892-27 Merck Index: 14,8627 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...