Zinc selenide, Optical Window, Diameter 12.7 mm (0.5 in), Thickness 2 mm (0.08 in)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc (II) selenide

045666-KS
AA45666-KSEA 344 USD
AA45666-KS
Zinc selenide, Optical Window, Diameter 12.7 mm (0.5 in), Thickness 2 mm (0.08 in)
Zinc selenide

Physical Form: Clear Aperture 85%, Surface Finish 60/40

Formula: SeZn
MW: 144.35 g/mol
Melting Pt: 1793 °C
Density: 5.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1,III
Merck Index: 12,10290

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR