Arsenic (III) sulfide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Arsenic trisulphide, Arsenic trisulfide, Disrsenic trisulfide, Disrsenic trisulphide
045616-06
045616-14
AA45616-06EA
91.3
USD
AA45616-06
AA45616-14
Arsenic (III) sulfide ≥99.9% (metals basis)
Arsenic (III) sulphide
Purity: 99.9%
CAS No.: 1303-33-9
Molecular Formula: As2S3
Formula Weight: 246.03
Physical Form: Powder
Density: 3.43
Melting Point: 300°
Boiling Point: 707°
Merck Reference: 14,806
UN No.: 1557
MDL No.: MFCD00003435
Formula:
As₂S₃ Boiling Pt: 707 °C Melting Pt: 300 °C Density: 3.43 Storage Temperature: Ambient |
MDL Number:
MFCD00003435 CAS Number: 1303-33-9 EINECS: 215-117-4 UN: 1557 ADR: 6.1,III Merck Index: 14,806 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...