Cobalt(II,III) oxide (∼3,4 - 4,5% CoO, ∼11.5 - 14.5% MoO) on alumina

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tricobalt tetraoxide, Cobaltic tetraoxide

045579-36 045579-22 045579-A4
AA45579-14EA 125.41 USD
AA45579-14 AA45579-36 AA45579-22 AA45579-A4
Cobalt(II,III) oxide (∼3,4 - 4,5% CoO, ∼11.5 - 14.5% MoO) on alumina
Cobalt (II,III) oxide

2.5 mm Trilobe extrudate


Physical Form: 2.5 mm Trilobe extrudate Appearance: Blue Odor: Odorless UN No.: 3077 MDL No.: MFCD01074445

Formula: Co₃O₄
MW: 240.8 g/mol
Melting Pt: 895
Storage Temperature: Ambient
MDL Number: MFCD00010939
CAS Number: 1308-06-1
UN: 3077
ADR: 9 III
Merck Index: 13,02454

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR