Palladium hydroxide (5% Pd) on carbon (Pearlman's catalyst) wetted_50%_Water, powder Type A403002-5

Supplier: Thermo Scientific Chemicals

045576-09 045576-04
AA45576-09EA 775 USD
AA45576-09 AA45576-04
Palladium hydroxide (5% Pd) on carbon (Pearlman's catalyst) wetted_50%_Water, powder Type A403002-5
Palladium hydroxide

CAS No.: 12135-22-7 Appearance: Black Odor: Odorless MDL No.: MFCD00064599

Formula: H₂O₂Pd
MW: 140.435 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064599
CAS Number: 12135-22-7
EINECS: 235-219-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR