Potassium tetrabromoplatinate(II) ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

045548-88 045548-ME
AA45548-88EA 738 USD
AA45548-88 AA45548-ME
Potassium tetrabromoplatinate(II) ≥99.9% (metals basis)
Potassium tetrabromoplatinate (II)

Purity: 99.9% CAS No.: 13826-94-3 Molecular Formula: Br4K2Pt Formula Weight: 592.90 Physical Form: Crystalline Melting Point: 300° MDL No.: MFCD00011376

Formula: Br₄K₂Pt
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00011376
CAS Number: 13826-94-3
EINECS: 237-536-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR