Gadolinium ≥99.9% (by rare eath metals basis), ingot

Supplier: Thermo Scientific Chemicals

045529-KG
AA45529-KGEA 184 USD
AA45529-KG
Gadolinium ≥99.9% (by rare eath metals basis), ingot
Gadolinium

Purity: 99.9% CAS No.: 7440-54-2 Physical Form: ≈30-50g/piece, irreg. shape, packaged under argon Sensitivity: Air and Moisture Sensitive MDL No.: MFCD00011022

Formula: Gd
MW: 157.25
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR