Rhenium ≥99.99% (metals basis), slug, Diameter 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals

045524-KT
AA45524-KTEA 349 USD
AA45524-KT AA45524-KU
Rhenium ≥99.99% (metals basis), slug, Diameter 12.7 mm (0.5 in)
Rhenium

Purity: 99.99% CAS No.: 7440-15-5 Physical Form: ≈6.35mm long, 7.5 +/- 2 grams Odor: Odorless MDL No.: MFCD00011195

Formula: Re
Boiling Pt: 5630 °C (1013 hPa)
Melting Pt: 3180 °C
Density: 21.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011195
CAS Number: 7440-15-5
EINECS: 231-124-5
Merck Index: 12,08343

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR