Ruthenium ≥99.9% (metals basis), cubes, Length 6 mm (0.24 in)

Supplier: Thermo Scientific Chemicals

045484-KF
AA45484-KFEA 1060 USD
AA45484-KF
Ruthenium ≥99.9% (metals basis), cubes, Length 6 mm (0.24 in)
Ruthenium

Purity: 99.9% CAS No.: 7440-18-8 Physical Form: ≈2-3g/cube Odor: Odorless MDL No.: MFCD00011207

Formula: Ru
MW: 101.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR