Lead(II) sulfide ≥99.995% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead sulfide, Lead sulphide

045462-09
AA45462-09EA 153 USD
AA45462-09
Lead(II) sulfide ≥99.995% (metals basis)
Lead (II) sulphide

Purity: 99.995% CAS No.: 1314-87-0 Molecular Formula: PbS Formula Weight: 239.25 Physical Form: Lump Density: 7.5 Refractive Index: 3.921 Odor: Odorless Melting Point: 1114° Merck Reference: 14,5421 UN No.: 3077 MDL No.: MFCD00016280

Formula: ̍PbS
MW: 239.27 g/mol
Boiling Pt: 1344
Melting Pt: 1114
Density: 7.5
Storage Temperature: Ambient
MDL Number: MFCD00016280
CAS Number: 1314-87-0
EINECS: 215-246-6
UN: 3077
ADR: 9 III
Merck Index: 14,5421

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR