Germanium, Optical Window, Diameter 25.4 mm (1.0 in), Thickness 2 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

045387-KS
AA45387-KSEA 358 USD
AA45387-KS
Germanium, Optical Window, Diameter 25.4 mm (1.0 in), Thickness 2 mm (0.08 in)
Germanium

CAS No.: 7440-56-4 Physical Form: Uncoated, Clear Aperture 85%, Surface Finish 40/20 MDL No.: MFCD00085310

Formula: Ge
Boiling Pt: 2830 °C (1013 hPa)
Melting Pt: 937.2 °C
Density: 5.323 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
Merck Index: 12,04415

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR