Thallium, Oil Based Standard Solution, Specpure®, Tl 1000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
045306-18
AA45306-18EA
78.68
USD
AA45306-18
Thallium, Oil Based Standard Solution, Specpure®, Tl 1000 µg/g, Thermo Scientific
Standards
Cationic Standards
Thallium Standards
Liquid
Physical Form: Liquid
MDL No.: MFCD01095325
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...