Thallium 203, 203Tl, Plasma Standard Solution, Specpure®, 203Tl 10µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
045288-AD
AA45288-ADEA
127
USD
AA45288-AD
Thallium 203, 203Tl, Plasma Standard Solution, Specpure®, 203Tl 10µg/ml, Thermo Scientific
Standards
Cationic Standards
Thallium Standards
Molecular Formula: Matrix: 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00084592
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...