Cadmium 106, 106Cd, Plasma Standard Solution, Specpure®, 106Cd 10 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
045274-AD
AA45274-ADEA
363
USD
AA45274-AD
Cadmium 106, 106Cd, Plasma Standard Solution, Specpure®, 106Cd 10 µg/ml, Thermo Scientific
Standards
Cationic Standards
Cadmium Standards
Molecular Formula: Matrix: 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00143885
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...