Boron 11, 11B, Plasma Standard solution, Specpure®, 11B 100 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
045273-AD
AA45273-ADEA
257
USD
AA45273-AD
Boron 11, 11B, Plasma Standard solution, Specpure®, 11B 100 µg/ml, Thermo Scientific
Standards
Cationic Standards
Boron Standards
Molecular Formula: Matrix: H2O
Physical Form: Liquid
MDL No.: MFCD00151272
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...