Zinc, Plasma Standard Solution, Specpure®, 10 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
045269-AE
AA45269-AEEA
80.1
USD
AA45269-AE
Zinc, Plasma Standard Solution, Specpure®, 10 µg/ml, Thermo Scientific
Standards
Cationic Standards
Zinc Standards
Molecular Formula: Zn in 2% HNO3
Physical Form: Liquid
MDL No.: MFCD00011291
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...