Terbium, Plasma Standard Solution, Specpure®, Tb 10 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
045266-AE
AA45266-AEEA
73
USD
AA45266-AE
Terbium, Plasma Standard Solution, Specpure®, Tb 10 µg/ml, Thermo Scientific
Standards
Cationic Standards
Terbium Standards
Molecular Formula: Tb4O7 in 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011256
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...