Tellurium, Plasma Standard Solution, Specpure®, Te 10 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
045265-AE
AA45265-AEEA
224
USD
AA45265-AE
Tellurium, Plasma Standard Solution, Specpure®, Te 10 µg/ml, Thermo Scientific
Standards
Cationic Standards
Tellurium Standards
Molecular Formula: Te in 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134062
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...