Silver, Plasma Standard Solution, Specpure®, Ag 10 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
045264-AE
AA45264-AEEA
75.72
USD
AA45264-AE
Silver, Plasma Standard Solution, Specpure®, Ag 10 µg/ml, Thermo Scientific
Standards
Cationic Standards
Silver Standards
Molecular Formula: Ag in 2% HNO3
Physical Form: Liquid
Density: ca 1.0
UN No.: 3264
MDL No.: MFCD00003397
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...