Nickel, Plasma Standard Solution, Specpure®, Ni 10 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
045261-AE
AA45261-AEEA
66.62
USD
AA45261-AE
Nickel, Plasma Standard Solution, Specpure®, Ni 10 µg/ml, Thermo Scientific
Standards
Cationic Standards
Nickel Standards
Molecular Formula: Ni in 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011137
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...