Copper, plasma standard solution, Specpure®, Cu 10 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
045253-AE
AA45253-AEEA
65.72
USD
AA45253-AE
Copper, plasma standard solution, Specpure®, Cu 10 µg/ml, Thermo Scientific
Standards
Cationic Standards
Copper Standards
Molecular Formula: Cu in 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010965
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...