Bismuth, Plasma Standard Solution, Specpure®, Bi 10µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
045249-AE
AA45249-AEEA
68.77
USD
AA45249-AE
Bismuth, Plasma Standard Solution, Specpure®, Bi 10µg/ml, Thermo Scientific
Standards
Cationic Standards
Bismuth Standards
Molecular Formula: Bi in 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134033
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...