Manganese ≥99.9998% (metals basis), pieces ≤ 10 mm, Ultrapure

Supplier: Thermo Scientific Chemicals

045130-14
AA45130-14EA 1164.25 USD
AA45130-14
Manganese ≥99.9998% (metals basis), pieces ≤ 10 mm, Ultrapure
Manganese

Purity: 99.9998% CAS No.: 7439-96-5 Molecular Formula: Mn Physical Form: Ampouled under vacuum Odor: Odorless

Formula: Mn
Storage Temperature: Ambient
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR