Indium ≥99.9% (metals basis), powder -100 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
045122-06
045122-14
AA45122-06EA
106.72
USD
AA45122-06
AA45122-14
Indium ≥99.9% (metals basis), powder -100 mesh
Indium
Purity: 99.9%
CAS No.: 7440-74-6
Molecular Formula: In
Formula Weight: 114.82
Physical Form: Powder
Density: 7.31
Appearance: Silver-grey
Odor: Odorless
Melting Point: 156.6°
Boiling Point: 2072°
UN No.: 3089
MDL No.: MFCD00134048
Formula:
In MW: 114.82 g/mol Boiling Pt: 2000 Melting Pt: 156 Density: 7.31 Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 UN: 3089 ADR: 4.1 III Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...