Magnesium, cubes ∼ 19 mm (0.75 in) each side, Length 19mm (0.75in)

Supplier: Thermo Scientific Chemicals

045119-KF
AA45119-KFEA 73.9 USD
AA45119-KF
Magnesium, cubes ∼ 19 mm (0.75 in) each side, Length 19mm (0.75in)
Magnesium

CAS No.: 7439-95-4 Physical Form: ≈10-15g/cube Appearance: Silver-colored Odor: Odorless MDL No.: MFCD00085308

Formula: Mg
MW: 24.3 g/mol
Boiling Pt: 1107
Melting Pt: 651
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1 III
Merck Index: 13,05675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR