Niobium ≥99.95% (metals basis), ingot/button, Diameter 25.4 mm (1.0 in), Thickness ∼11 mm (0.45 in)

Supplier: Thermo Scientific Chemicals

045113-KF
AA45113-KFEA 148 USD
AA45113-KF
Niobium ≥99.95% (metals basis), ingot/button, Diameter 25.4 mm (1.0 in), Thickness ∼11 mm (0.45 in)
Niobium

Purity: 99.95% CAS No.: 7440-03-1 Physical Form: ≈50g/piece Appearance: Silver-grey Odor: Odorless MDL No.: MFCD00011126

Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR