Indium ≥99.99% (metals basis), ingot/button, Diameter ∼31 mm (1.2 in), Thickness 5.3 mm (0.21 in)

Supplier: Thermo Scientific Chemicals

045111-KF
AA45111-KFEA 150 USD
AA45111-KF
Indium ≥99.99% (metals basis), ingot/button, Diameter ∼31 mm (1.2 in), Thickness 5.3 mm (0.21 in)
Indium

Purity: 99.99% CAS No.: 7440-74-6 Physical Form: ≈28g/piece Appearance: Silver Odor: Odorless MDL No.: MFCD00134048

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR