Tellurium ≥99.9% (metals basis), ingot/button, Diameter ∼38 mm (1.5 in), Thickness 6 mm (0.24 in)

Supplier: Thermo Scientific Chemicals
Danger

045108-KF
AA45108-KFEA 148 USD
AA45108-KF
Tellurium ≥99.9% (metals basis), ingot/button, Diameter ∼38 mm (1.5 in), Thickness 6 mm (0.24 in)
Tellurium

Purity: 99.9% CAS No.: 13494-80-9 Physical Form: ≈30g/piece Appearance: Silver Odor: Odorless MDL No.: MFCD00134062

Formula: Te
MW: 127.6 g/mol
Boiling Pt: 990
Melting Pt: 450
Density: 8.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3284
ADR: 6.1 III
Merck Index: 12,09273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR